Skip to Content
Merck
Silicon sphere, 0.48 mm diameter, 20 pcs, tolerance +/-2.5um

Silicon

Silicon sphere, 0.48 mm diameter, 20 pcs, tolerance +/-2.5um
Synonym(s):
Silicon
Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09

Products

Shipping
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Quality Segment
Showing 1-20 of 80
Page 1 of 4
Compare
Product Number
Description
Pricing
Page 1 of 4