Sesamol
Synonym(s):
3,4-(Methylenedioxy)phenol, 5-Benzodioxolol
Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing