Skip to Content
Merck
(S)-(+)-α-Methoxyphenylacetic acid 99%

(S)-(+)-α-Methoxyphenylacetic acid

(S)-(+)-α-Methoxyphenylacetic acid 99%
Synonym(s):
O-Methyl-L-mandelic acid
Linear Formula:
C6H5CH(OCH3)CO2H
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing