Skip to Content
Merck
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione 97%

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione 97%
Synonym(s):
(+)-5,6,7,8-Tetrahydro-8-methylindan-1,5-dione, (+)-7,7a-Dihydro-7α, β-methyl-1,5(6H)-indandione, (S)-(+)-Hajos-Parrish diketone, (S)-Hajos dione, (S)-Hajos ketone, (S)-Hajos-Wiechert ketone
Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing