![(S)-β-Amino-3-[2-(dimethylamino)ethyl]-1H-indole-5-propanol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/925/155/55c2a5f1-92e8-4562-9f64-93f90a17b45b/640/55c2a5f1-92e8-4562-9f64-93f90a17b45b.jpg)
(S)-β-Amino-3-[2-(dimethylamino)ethyl]-1H-indole-5-propanol
![(S)-β-Amino-3-[2-(dimethylamino)ethyl]-1H-indole-5-propanol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/925/155/55c2a5f1-92e8-4562-9f64-93f90a17b45b/640/55c2a5f1-92e8-4562-9f64-93f90a17b45b.jpg)
Synonym(s):
(S)-2-Amino-3-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}propan-1-ol
Empirical Formula (Hill Notation):
C15H23N3O
CAS Number:
Molecular Weight:
261.36
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing