Saxagliptin monohydrate
Synonym(s):
Saxagliptin hydrate, (S)-3-Hydroxyadamantylglycine-L-cis-4,5-methanoprolinenitrile hydrate, BMS 477118, BMS 477118-11, BMS-477118, BMS-477118-11, BMS477118, BMS477118-11
Empirical Formula (Hill Notation):
C18H25N3O2 · H2O
CAS Number:
Molecular Weight:
333.43
Products
Brand
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing