Skip to Content
Merck
Sarcosine ethyl ester hydrochloride 99%

Sarcosine ethyl ester hydrochloride

Sarcosine ethyl ester hydrochloride 99%
Synonym(s):
Ethyl sarcosinate hydrochloride
Linear Formula:
CH3NHCH2COOC2H5 · HCl
CAS Number:
Molecular Weight:
153.61
EC Number:

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing