Skip to Content
Merck
Sarafotoxin S6c ≥97% (HPLC)

Sarafotoxin S6c

Sarafotoxin S6c ≥97% (HPLC)
Synonym(s):
SRTX-c
Empirical Formula (Hill Notation):
C103H147N27O37S5
CAS Number:
Molecular Weight:
2515.75

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing