Skip to Content
Merck
Salvinorin A ≥98% (HPLC), solid

Salvinorin A

Salvinorin A ≥98% (HPLC), solid
Synonym(s):
Salvinorin A, (2S,4aR,6aR,7R,9S,10aS,10bR)-9-(acetyloxy)-2-(3-furanyl)dodechydro-6a,10b-dimethyl-4,10-dioxo-2H-naphtho[2,1-c]pyran-7-carboxylic acid methyl ester
Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing