Skip to Content
Merck
(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥97%

(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine

(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥97%
Synonym(s):
(2S)-1-[(1S)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS), (S)-(R)-Josiphos, Josiphos SL-J001-2
Empirical Formula (Hill Notation):
C36H44FeP2 · C2H5OH
CAS Number:
Molecular Weight:
640.60

Products

Shipping
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing