![Rubusoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/402/013/d1d2f159-aca9-4f56-94a1-2567d348cc6a/640/d1d2f159-aca9-4f56-94a1-2567d348cc6a.png)
Rubusoside
![Rubusoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/402/013/d1d2f159-aca9-4f56-94a1-2567d348cc6a/640/d1d2f159-aca9-4f56-94a1-2567d348cc6a.png)
Synonym(s):
(4α)-13-(β-D-Glucopyranosyloxy)-kaur-16-en-18-oic acid β-D-glucopyranosyl ester, (4α)-13-(β-D-glucopyranosyloxy)kaur-16-en-18-oic acid β-D-glucopyranosyl ester
Empirical Formula (Hill Notation):
C32H50O13
CAS Number:
Molecular Weight:
642.73
Products
Shipping
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing