Skip to Content
Merck
vibrant-m

Rotigotine

vibrant-m
Synonym(s):
(6S)-5,6,7,8-Tetrahydro-6-[propyl[2-(2-thienyl)ethyl]amino]-1-naphthalenol, (S)-5,6,7,8-Tetrahydro-6-(propyl(2-(2-thienyl)ethyl)amino)-1-naphthol
Empirical Formula (Hill Notation):
C19H25NOS
Molecular Weight:
315.47

Products

Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2