Skip to Content
Merck
Ropinirole hydrochloride European Pharmacopoeia (EP) Reference Standard

Ropinirole hydrochloride

Ropinirole hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-[2-(Dipropylamino)ethyl]-1,3-dihydro-2H-indol-2-one hydrochloride, SKF 101468 hydrochloride, Ropinirole hydrochloride
Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84

Products

Shipping
Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique