
Rifamycin B

Empirical Formula (Hill Notation):
C39H49NO14
CAS Number:
Molecular Weight:
755.80
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing