Rhodium(II) trifluoroacetate dimer
Synonym(s):
Trifluoroacetic acid rhodium(II) salt dimer
Linear Formula:
[(CF3COO)2Rh]2
CAS Number:
Molecular Weight:
657.87
Products
Shipping
Brand
Formula Weight
Physical Form
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing