Skip to Content
Merck
(R)-(−)-Glycidyl methyl ether 97%

(R)-(−)-Glycidyl methyl ether

(R)-(−)-Glycidyl methyl ether 97%
Synonym(s):
(R)-(−)-2-(Methoxymethyl)oxirane, (R)-(−)-Methyl glycidyl ether
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing