Skip to Content
Merck
vibrant-m

Residual Solvent Class 2 - Chlorobenzene

vibrant-m
Synonym(s):
Chlorobenzene solution
Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56

Products

Shipping
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1