Skip to Content
Merck
vibrant-m

Residual Solvent Class 1 - Benzene

vibrant-m
Synonym(s):
Benzene solution
Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11

Products

Shipping
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1