Skip to Content
Merck
vibrant-m

Residual Solvent Class 1 - 1,1,1-Trichloroethane

vibrant-m
Synonym(s):
1,1,1-Trichloroethane solution, Methyl chloroform
CAS Number:

Products

Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1