(R)-(+)-Bornylamine
Synonym(s):
(+)-Bornanamine, endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine
Empirical Formula (Hill Notation):
C10H19N
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing