Skip to Content
Merck
Rafoxanide PESTANAL®, analytical standard

Rafoxanide

Rafoxanide PESTANAL®, analytical standard
Synonym(s):
3′-Chloro-4′-(4-chlorophenoxy)-3,5-diiodosalicylanilide
Empirical Formula (Hill Notation):
C19H11Cl2I2NO3
CAS Number:
Molecular Weight:
626.01
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing