Skip to Content
Merck
(R)-(−)-1-(1-Naphthyl)ethyl isocyanate 98%

(R)-(−)-1-(1-Naphthyl)ethyl isocyanate

(R)-(−)-1-(1-Naphthyl)ethyl isocyanate 98%
Synonym(s):
(R)-NEI
Linear Formula:
C10H7CH(CH3)NCO
CAS Number:
Molecular Weight:
197.23
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing