
p-Xylene-bis(N-pyridinium bromide)

Synonym(s):
DPX
Empirical Formula (Hill Notation):
C18H18Br2N2
CAS Number:
Molecular Weight:
422.16
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing