PTX-11 solution
Synonym(s):
Pectenotoxin 11 solution
Empirical Formula (Hill Notation):
C47H70O15
CAS Number:
Molecular Weight:
875.05
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1