Protease from Bacillus licheniformis
Synonym(s):
Proteinase from Bacillus licheniformis, Subtilisin A
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Quality Segment
Application
Showing 1-4 of 4
Compare
Product Number
Description
Pricing