Skip to Content
Merck
Propylamine analytical standard

Propylamine

Propylamine analytical standard
Synonym(s):
1-Aminopropane, Propylamine
Linear Formula:
CH3CH2CH2NH2
CAS Number:
Molecular Weight:
59.11
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing