
Potassium ionophore III

Synonym(s):
2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5′-nitro(benzo-15-crown-5)-4′-yl]carbamate], BME 44
Empirical Formula (Hill Notation):
C46H70N4O18
CAS Number:
Molecular Weight:
967.06
Products
Shipping
Product Category
Formula Weight
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing