Skip to Content
Merck
Potassium hexafluorotitanate(IV)

Potassium hexafluorotitanate(IV)

Potassium hexafluorotitanate(IV)
Synonym(s):
Dipotassium hexafluorotitanate, Titanium potassium hexafluoride
Linear Formula:
K2(TiF6)
CAS Number:
Molecular Weight:
240.05
EC Number:

Products

Shipping
Formula Weight
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing