Poly(styrene-co-divinylbenzene) Polymeric Adsorbent
Synonym(s):
Polystyrene crosslinked with divinylbenzene
Linear Formula:
[CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y
CAS Number:
Products
Shipping
Product Category
Brand
Physical Form
Separation Technique
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing