Skip to Content
Merck
Polyetherimide melt index 18 g/10 min (337 °C/6.6kg)

Polyetherimide

Polyetherimide melt index 18 g/10 min (337 °C/6.6kg)
Synonym(s):
Poly(bisphenol A-co-4-nitrophthalic anhydride-co-1,3-phenylenediamine), PEI, Poly(bisphenol A anhydride-co-1,3-phenylenediamine), Poly(bisphenol A dianhydride-co-1,3-phenylenediamine), Poly(ether imide), Poly-[2,2′bis(4-(3,4-dicarboxyphenoxy)phenylpropane)-1,3-phenylene bisimide], Polyetherimide
Linear Formula:
(C39H30N2O6)n
CAS Number:

Products

Shipping
Brand
Physical Form
Quality Segment
Polymer Type
Showing 1-2 of 2