
Plastic additive 8

Synonym(s):
Dioctadecyl disulfide
Empirical Formula (Hill Notation):
C36H74S2
CAS Number:
Molecular Weight:
571.10
Products
Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2