Skip to Content
Merck
Phenylselenyl bromide 98%

Phenylselenyl bromide

Phenylselenyl bromide 98%
Synonym(s):
Benzeneselenenyl bromide, Benzeneselenyl bromide
Linear Formula:
C6H5SeBr
CAS Number:
Molecular Weight:
235.97
EC Number:

Products

Shipping
Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing