Skip to Content
Merck
Phenylephrine hydrochloride British Pharmacopoeia (BP) Reference Standard

Phenylephrine Hydrochloride

Phenylephrine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
(R)-(−)-Phenylephrine hydrochloride, (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride
Linear Formula:
HOC6H4CH(CH2NHCH3)OH·HCl
CAS Number:
Molecular Weight:
203.67
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique