Phenelzine sulfate
Synonym(s):
Phenelzine sulfate salt, Phenelzine hydrogen sulphate
Empirical Formula (Hill Notation):
C8H12N2 · H2O4S
CAS Number:
Molecular Weight:
234.27
Products
Shipping
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing