Pentobarbital sodium
Synonym(s):
Pentobarbital sodium salt, 5-Ethyl-5-(1-methylbutyl)-2,4,6-trioxohexahydropyrimidine, Nembutal
Empirical Formula (Hill Notation):
C11H17N2NaO3
CAS Number:
Molecular Weight:
248.25
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing