Skip to Content
Merck
PCB No 29 analytical standard

PCB No 29

PCB No 29 analytical standard
Synonym(s):
2,4,5-Trichlorobiphenyl
Empirical Formula (Hill Notation):
C12H7Cl3
CAS Number:
Molecular Weight:
257.54
EC Number:

Products

Shipping
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing