
Paraformaldehyde-d2

Synonym(s):
Deuterated paraformaldehyde, Dideuteroparaformaldehyde, Formaldehyde-d2 polymer, Polyoxymethylene-D2
Linear Formula:
(CD2O)n
CAS Number:
Molecular Weight:
32.04
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing