Skip to Content
Merck
Palatinose hydrate ≥99% (GC)

Palatinose hydrate

Palatinose hydrate ≥99% (GC)
Synonym(s):
6-O-α-D-Glucopyranosyl-D-fructose, Isomaltulose hydrate
Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing