![Oxcarbazepine Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/408/386/2950994b-6733-46c0-b776-c34f4f067c67/640/2950994b-6733-46c0-b776-c34f4f067c67.jpg)
Oxcarbazepine Related Compound A
![Oxcarbazepine Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/408/386/2950994b-6733-46c0-b776-c34f4f067c67/640/2950994b-6733-46c0-b776-c34f4f067c67.jpg)
Synonym(s):
N-Formyl-10-oxo-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide
Empirical Formula (Hill Notation):
C16H12N2O3
CAS Number:
Molecular Weight:
280.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2