Oxazepam Related Compound A
Synonym(s):
7-Chloro-4,5-dihydro-5-phenyl-1H-1,4-benzodiazepine-2,3-dione
Empirical Formula (Hill Notation):
C15H11ClN2O2
CAS Number:
Molecular Weight:
286.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing