Skip to Content
Merck
Oxacyclotridecan-2-one 98%

Oxacyclotridecan-2-one

Oxacyclotridecan-2-one 98%
Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing