Skip to Content
Merck
O-Tricyclo[5.2.1.02,6]dec-9-yl dithiocarbonate potassium salt ≥95%, solid

O-Tricyclo[5.2.1.02,6]dec-9-yl dithiocarbonate potassium salt

O-Tricyclo[5.2.1.02,6]dec-9-yl dithiocarbonate potassium salt ≥95%, solid
Synonym(s):
D609
Empirical Formula (Hill Notation):
C11H15KOS2
CAS Number:
Molecular Weight:
266.46
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing