Ondansetron impurity D
Synonym(s):
9-Methyl-3-methylene-1,2,3,9-tetrahydro-4H-carbazol-4-one, 1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-one
Empirical Formula (Hill Notation):
C14H13NO
CAS Number:
Molecular Weight:
211.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing