Skip to Content
Merck
Ofurace PESTANAL®, analytical standard

Ofurace

Ofurace PESTANAL®, analytical standard
Synonym(s):
2-Chloro-N-(2,6-dimethylphenyl)-N-(tetrahydro-2-oxo-3-furanyl)acetamide
Empirical Formula (Hill Notation):
C14H16ClNO3
CAS Number:
Molecular Weight:
281.73
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing