Skip to Content
Merck
o-Flutamide certified reference material, pharmaceutical secondary standard

o-Flutamide

o-Flutamide certified reference material, pharmaceutical secondary standard
Synonym(s):
2-Methyl-N-[6-nitro-3-(trifluoromethyl)phenyl]propanamide
Empirical Formula (Hill Notation):
C11H11F3N2O3
CAS Number:
Molecular Weight:
276.21

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade