Skip to Content
Merck
O-Desmethylvenlafaxine solution 1.0 mg/mL in methanol, certified reference material, Cerilliant®

O-Desmethylvenlafaxine solution

O-Desmethylvenlafaxine solution 1.0 mg/mL in methanol, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C16H25NO2
CAS Number:
Molecular Weight:
263.38

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique