
N-tert-Butyltrimethylsilylamine

Synonym(s):
N-(Trimethylsilyl)-tert-butylamine
Linear Formula:
(CH3)3SiNHC(CH3)3
CAS Number:
Molecular Weight:
145.32
Products
Shipping
Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing