![Noradrenaline impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/236/942/d2e041db-f14d-4c43-8820-2f0d8e2b64dc/640/d2e041db-f14d-4c43-8820-2f0d8e2b64dc.png)
Noradrenaline impurity E
![Noradrenaline impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/236/942/d2e041db-f14d-4c43-8820-2f0d8e2b64dc/640/d2e041db-f14d-4c43-8820-2f0d8e2b64dc.png)
Synonym(s):
2-Chloro-3′,4′-dihydroxyacetophenone, 4-(Chloroacetyl)catechol
Linear Formula:
(HO)2C6H3COCH2Cl
CAS Number:
Molecular Weight:
186.59
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing