Nobiletin
Synonym(s):
2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-1-benzopyran-4-one, 3′,4′,5,6,7,8-Hexamethoxyflavone
Empirical Formula (Hill Notation):
C21H22O8
CAS Number:
Molecular Weight:
402.39
Products
Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing