Skip to Content
Merck
N-Nonanoyl-N-methylglucamine ≥98%

N-Nonanoyl-N-methylglucamine

N-Nonanoyl-N-methylglucamine ≥98%
Synonym(s):
N-(D-Glucityl)-N-methylnonanamide, N-Methyl-N-nonanoyl-D-glucamine, MEGA-9
Empirical Formula (Hill Notation):
C16H33NO6
CAS Number:
Molecular Weight:
335.44

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing